4-{[5-(Trifluoromethyl)-2-pyridinyl]sulfanyl}aniline ≥95%

Supplier: Matrix Scientific

054923-500MG 054923-1G
101909-078EA 556.92 USD
101909-078 101909-080
4-{[5-(Trifluoromethyl)-2-pyridinyl]sulfanyl}aniline ≥95%
4-{[5-(Trifluoromethyl)-2-pyridinyl]sulfanyl}aniline

Hazard: Irritant

Formula: C₁₂H₉F₃N₂S
MW: 270.28 g/mol
Melting Pt: 61…63 °C
MDL Number: MFCD11132350
CAS Number: 1019391-05-9

Order Now

Specification Test Results

Melting point 61-63°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR