tert-Butyl-4-(2-ethoxy-2-oxoethylidene)tetrahydro-1(2H)-pyridinecarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
4-(2-Ethoxy-2-oxoethylidene)piperidine-1-carboxylic acid tert-Butyl ester, Ethyl [1-(tert-Butoxycarbonyl)piperidin-4-ylidene]acetate, tert-Butyl 4-(2-Ethoxy-2-oxoethylidene)piperidine-1-carboxylate, (1-Boc-piperidin-4-ylidene)acetic acid ethyl ester, [1-(tert-Butoxycarbonyl)piperidin-4-ylidene]acetic acid ethyl ester, Ethyl (1-Boc-piperidin-4-ylidene)acetate
054921-500MG
054921-1G
054921-5G
101909-066EA
200
USD
101909-066
101909-068
101909-070
tert-Butyl-4-(2-ethoxy-2-oxoethylidene)tetrahydro-1(2H)-pyridinecarboxylate ≥95%
tert-Butyl-4-(2-ethoxy-2-oxoethylidene)tetrahydro-1(2H)-pyridinecarboxylate
Hazard: Irritant
Formula:
C₁₄H₂₃NO₄ MW: 269.34 g/mol Melting Pt: 88…89 °C |
MDL Number:
MFCD12912655 CAS Number: 135716-08-4 |
Specification Test Results
Melting point | 88-89°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...