tert-Butyl-4-amino-4-cyanotetrahydro-1(2H)-pyridinecarboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 4-amino-4-cyanopiperidine-1-carboxylate, 4-Amino-4-cyanopiperidine, N1-BOC protected

054920-500MG 054920-1G
101909-062EA 184.62 USD
101909-062 101909-064
tert-Butyl-4-amino-4-cyanotetrahydro-1(2H)-pyridinecarboxylate ≥97%
tert-Butyl-4-amino-4-cyanotetrahydro-1(2H)-pyridinecarboxylate

Hazard: Irritant

Formula: C₁₁H₁₉N₃O₂
MW: 225.29 g/mol
Melting Pt: 91…92 °C
MDL Number: MFCD08234564
CAS Number: 331281-25-5

Order Now

Specification Test Results

Melting point 91-92°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR