8-Phenyl-1-thia-4-azaspiro[4.5]decane-8-carbonitrile hydrochloride ≥95%

Supplier: Matrix Scientific

054919-500MG 054919-1G
101909-058EA 278.46 USD
101909-058 101909-060
8-Phenyl-1-thia-4-azaspiro[4.5]decane-8-carbonitrile hydrochloride ≥95%
8-Phenyl-1-thia-4-azaspiro[4.5]decane-8-carbonitrile hydrochloride

Hazard: Irritant

Formula: C₁₅H₁₉ClN₂S
Melting Pt: 223…226 °C
MDL Number: MFCD16140372
CAS Number: 1221792-40-0

Order Now

Specification Test Results

Melting point 223-226°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR