8-Phenyl-1-thia-4-azaspiro[4.5]decane-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

054916-500MG 054916-1G 054916-5G
101909-044EA 278.46 USD
101909-044 101909-046 101909-048
8-Phenyl-1-thia-4-azaspiro[4.5]decane-3-carboxylic acid ≥95%
8-Phenyl-1-thia-4-azaspiro[4.5]decane-3-carboxylic acid

Hazard: Irritant

Formula: C₁₅H₁₉NO₂S
Melting Pt: 207 °C (dec.)
MDL Number: MFCD02648392
CAS Number: 367928-68-5

Order Now

Specification Test Results

Melting point 207°C(dec)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR