8-Methyl-1-thia-4-azaspiro[4.5]decane-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

054913-500MG 054913-1G
101909-032EA 556.92 USD
101909-032 101909-034
8-Methyl-1-thia-4-azaspiro[4.5]decane-3-carboxylic acid ≥95%
8-Methyl-1-thia-4-azaspiro[4.5]decane-3-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₁₇NO₂S
Melting Pt: 156…158 °C
MDL Number: MFCD06633105
CAS Number: 1037010-80-2

Order Now

Specification Test Results

Melting point 156-158°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR