Methyl-5-methoxy-2-oxo-2,3-dihydro-1,3-benzoxazole-7-carboxylate ≥95%

Supplier: Matrix Scientific

054904-500MG 054904-1G
101909-002EA 529.23 USD
101909-002 101909-004
Methyl-5-methoxy-2-oxo-2,3-dihydro-1,3-benzoxazole-7-carboxylate ≥95%
Methyl-5-methoxy-2-oxo-2,3-dihydro-1,3-benzoxazole-7-carboxylate

Hazard: Irritant

Formula: C₁₀H₉NO₅
Melting Pt: 244…246 °C
MDL Number: MFCD16140364
CAS Number: 1221791-71-4

Order Now

Specification Test Results

Melting point 244-246°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR