Methyl-7-methoxy-2-oxo-2,3-dihydro-1,3-benzoxazole-5-carboxylate ≥95%

Supplier: Matrix Scientific

054903-500MG 054903-1G
101908-998EA 556.92 USD
101908-998 101909-000
Methyl-7-methoxy-2-oxo-2,3-dihydro-1,3-benzoxazole-5-carboxylate ≥95%
Methyl-7-methoxy-2-oxo-2,3-dihydro-1,3-benzoxazole-5-carboxylate

Hazard: Irritant

Formula: C₁₀H₉NO₅
Melting Pt: 211…212 °C
MDL Number: MFCD16140363
CAS Number: 1221792-78-4

Order Now

Specification Test Results

Melting point 211-212°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR