Methyl-5-methoxy-2-methyl-1,3-benzoxazole-7-carboxylate ≥95%

Supplier: Matrix Scientific

054902-500MG 054902-1G
101908-994EA 556.92 USD
101908-994 101908-996
Methyl-5-methoxy-2-methyl-1,3-benzoxazole-7-carboxylate ≥95%
Methyl-5-methoxy-2-methyl-1,3-benzoxazole-7-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₁NO₄
MW: 221.21 g/mol
Melting Pt: 119…121 °C
MDL Number: MFCD16140362
CAS Number: 1221792-76-2

Order Now

Specification Test Results

Melting point 119-121°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR