Methyl-5-methoxy-1,3-benzoxazole-7-carboxylate ≥95%
Supplier: Matrix Scientific
054901-500MG
054901-1G
101908-990EA
556.92
USD
101908-990
101908-992
Methyl-5-methoxy-1,3-benzoxazole-7-carboxylate ≥95%
Methyl-5-methoxy-1,3-benzoxazole-7-carboxylate
Hazard: Irritant
Formula:
C₁₀H₉NO₄ Melting Pt: 142…144 °C |
MDL Number:
MFCD16140361 CAS Number: 1221792-16-0 |
Specification Test Results
Melting point | 142-144°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...