9,9-Bis(4-hydroxyphenyl)fluorene ≥99%

Supplier: Matrix Scientific

Synonyms: 4,4'-(9-Fluorenylidene)diphenol, 4,4'-(9H-Fluorene-9,9-diyl)diphenol

048732-25G 048732-100G
101885-044EA 44.62 USD
101885-044 101885-046
9,9-Bis(4-hydroxyphenyl)fluorene ≥99%
9,9-Bis(4-hydroxyphenyl)fluorene

Hazard: Irritant

Formula: C₂₅H₁₈O₂
MW: 350.42 g/mol
Melting Pt: 227 °C
MDL Number: MFCD00191392
CAS Number: 3236-71-3

Order Now

Specification Test Results

Melting point 227°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR