tert-Butyl-4-[(dibenzylamino)methyl]-4-hydroxytetrahydro-1(2H)-pyridinecarboxylate ≥95%

Supplier: Matrix Scientific

048719-500MG 048719-1G
101885-004EA 747.69 USD
101885-004 101885-006
tert-Butyl-4-[(dibenzylamino)methyl]-4-hydroxytetrahydro-1(2H)-pyridinecarboxylate ≥95%
tert-Butyl-4-[(dibenzylamino)methyl]-4-hydroxytetrahydro-1(2H)-pyridinecarboxylate

Hazard: Irritant

Formula: C₂₅H₃₄N₂O₃
MW: 410.56 g/mol
Melting Pt: 92…93 °C
MDL Number: MFCD13193119
CAS Number: 1192263-81-2

Order Now

Specification Test Results

Melting point 92-93°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR