(4-Nitrophenyl)(tetrahydro-1H-pyrazol-1-yl)methanone ≥95%

Supplier: Matrix Scientific

048715-500MG 048715-1G
101884-990EA 921.54 USD
101884-990 101884-992
(4-Nitrophenyl)(tetrahydro-1H-pyrazol-1-yl)methanone ≥95%
(4-Nitrophenyl)(tetrahydro-1H-pyrazol-1-yl)methanone

Hazard: Irritant

Formula: C₁₀H₁₁N₃O₃
Melting Pt: 104…106 °C
MDL Number: MFCD13193115
CAS Number: 900189-04-0

Order Now

Specification Test Results

Melting point 104-106°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR