Methyl-1H-pyrrolo[3,2-c]pyridin-4-ylether ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Methoxy-1H-pyrrolo[3,2-c]pyridine

048713-500MG 048713-1G
101884-982EA 320 USD
101884-982 101884-984
Methyl-1H-pyrrolo[3,2-c]pyridin-4-ylether ≥95%
Methyl-1H-pyrrolo[3,2-c]pyridin-4-ylether

Hazard: Irritant

Formula: C₈H₈N₂O
MW: 148.17 g/mol
Melting Pt: 90…91 °C
MDL Number: MFCD08690141
CAS Number: 944900-76-9

Order Now

Specification Test Results

Melting point 90-91°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR