5-Methyl[1,6]naphthyridin-2(1H)-one ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Methyl-1,2-dihydro-1,6-naphthyridin-2-one

048712-500MG 048712-1G
101884-978EA 747.69 USD
101884-978 101884-980
5-Methyl[1,6]naphthyridin-2(1H)-one ≥95%
5-Methyl[1,6]naphthyridin-2(1H)-one

Hazard: Irritant

Formula: C₉H₈N₂O
MW: 160.18 g/mol
Melting Pt: 246…248 °C
MDL Number: MFCD00867139
CAS Number: 88296-61-1

Order Now

Specification Test Results

Melting point 246-248°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR