Methyl-1-methyl-1H-1,3-benzimidazole-5-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 1-methyl-1H-benzo[d]imidazole-5-carboxylate

048711-500MG 048711-1G
101884-974EA 921.54 USD
101884-974 101884-976
Methyl-1-methyl-1H-1,3-benzimidazole-5-carboxylate ≥95%
Methyl-1-methyl-1H-1,3-benzimidazole-5-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₀N₂O₂
MW: 190.2 g/mol
Melting Pt: 109…111 °C
MDL Number: MFCD04492113
CAS Number: 131020-36-5

Order Now

Specification Test Results

Melting point 109-111°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR