Methyl-3-hydroxy-4-iodobenzoate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 3-hydroxy-4-iodobenzoate
048709-1G
048709-5G
101884-966EA
44.62
USD
101884-966
101884-968
Methyl-3-hydroxy-4-iodobenzoate ≥95%
Methyl-3-hydroxy-4-iodobenzoate
Hazard: Irritant
Formula:
C₈H₇IO₃ Melting Pt: 53…54 °C |
MDL Number:
MFCD06203960 CAS Number: 157942-12-6 |
Specification Test Results
Melting point | 53-54°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...