Methyl-4,6-dichloro-5-nitronicotinate ≥95%
Supplier: Matrix Scientific
048705-500MG
048705-1G
048705-5G
101884-948EA
393.85
USD
101884-948
101884-950
101884-952
Methyl-4,6-dichloro-5-nitronicotinate ≥95%
Methyl-4,6-dichloro-5-nitronicotinate
Hazard: Irritant
Formula:
C₇H₄Cl₂N₂O₄ MW: 251.03 g/mol Melting Pt: 65…66 °C |
MDL Number:
MFCD13193110 CAS Number: 89247-05-2 |
Specification Test Results
Melting point | 65-66°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...