Methyl-3,6-dibromo-2-hydroxy-1-naphthoate ≥95%

Supplier: Matrix Scientific

048704-500MG 048704-1G 048704-5G
101884-942EA 278.46 USD
101884-942 101884-944 101884-946
Methyl-3,6-dibromo-2-hydroxy-1-naphthoate ≥95%
Methyl-3,6-dibromo-2-hydroxy-1-naphthoate

Hazard: Irritant

Formula: C₁₂H₈Br₂O₃
Melting Pt: 176.5…177.5 °C
MDL Number: MFCD13193109
CAS Number: 1192263-94-7

Order Now

Specification Test Results

Melting point 176.5-177.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR