Methyl-4-chloro-1H-pyrrole-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-chloro-1H-pyrrole-2-carboxylate

048703-500MG 048703-1G
101884-938EA 615.38 USD
101884-938 101884-940
Methyl-4-chloro-1H-pyrrole-2-carboxylate ≥95%
Methyl-4-chloro-1H-pyrrole-2-carboxylate

Hazard: Irritant

Formula: C₆H₆ClNO₂
MW: 159.57 g/mol
Melting Pt: 100…101 °C
MDL Number: MFCD11176523
CAS Number: 1194-96-3

Order Now

Specification Test Results

Melting point 100-101°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR