Methyl-4-bromosalicylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 4-bromo-2-hydroxybenzoate
048701-1G
048701-5G
101884-930EA
60
USD
101884-930
101884-932
Methyl-4-bromosalicylate ≥95%
Methyl 4-bromosalicylate
Hazard: Irritant
Formula:
C₈H₇BrO₃ MW: 231.0434 g/mol Melting Pt: 41-42°C |
MDL Number:
MFCD07780736 CAS Number: 22717-56-2 |
Specification Test Results
Melting point | 41-42°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...