1-Methyl-1H-1,3-benzimidazole-5-carboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
1-Methyl-1H-benzo[d]imidazole-5-carboxylic acid
048700-500MG
048700-1G
101884-924EA
75.38
USD
101884-924
101884-926
1-Methyl-1H-1,3-benzimidazole-5-carboxylic acid ≥95%
1-Methyl-1H-1,3-benzimidazole-5-carboxylic acid
Hazard: Irritant
Formula:
C₉H₈N₂O₂ MW: 176.18 g/mol Melting Pt: 293…295 °C |
MDL Number:
MFCD06809665 CAS Number: 53484-17-6 |
Specification Test Results
Melting point | 293-295°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...