Methyl-4-amino-1H-pyrrole-2-carboxylate hydrochloride ≥95%

Supplier: Matrix Scientific

048698-500MG 048698-1G
101884-918EA 921.54 USD
101884-918 101884-920
Methyl-4-amino-1H-pyrrole-2-carboxylate hydrochloride ≥95%
Methyl-4-amino-1H-pyrrole-2-carboxylate hydrochloride

Hazard: Irritant

Formula: C₆H₉ClN₂O₂
Melting Pt: 231 °C (dec.)
MDL Number: MFCD09971538
CAS Number: 1171639-98-7

Order Now

Specification Test Results

Melting point 231(dec)°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR