Methyl-4-amino-3-bromo-5-methoxybenzenecarboxylate ≥95%
Supplier: Matrix Scientific
048696-500MG
048696-1G
048696-5G
101884-908EA
223.08
USD
101884-908
101884-910
101884-912
Methyl-4-amino-3-bromo-5-methoxybenzenecarboxylate ≥95%
Methyl-4-amino-3-bromo-5-methoxybenzenecarboxylate
Hazard: Irritant
Formula:
C₉H₁₀BrNO₃ Melting Pt: 91…93 °C |
MDL Number:
MFCD13193106 CAS Number: 762292-57-9 |
Specification Test Results
Melting point | 91-93°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...