Ethyl-2-phenoxy-5-(trifluoromethyl)nicotinate ≥95%

Supplier: Matrix Scientific

048693-500MG 048693-1G
101884-896EA 500 USD
101884-896 101884-898
Ethyl-2-phenoxy-5-(trifluoromethyl)nicotinate ≥95%
Ethyl-2-phenoxy-5-(trifluoromethyl)nicotinate

Hazard: Irritant

Formula: C₁₅H₁₂F₃NO₃
MW: 311.26 g/mol
Melting Pt: 48…50 °C
MDL Number: MFCD13193104
CAS Number: 1192263-87-8

Order Now

Specification Test Results

Melting point 48-50°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR