Ethyl-2-cyano-5-(trifluoromethyl)nicotinate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 2-cyano-5-(trifluoromethyl)nicotinate
048692-500MG
048692-1G
101884-892EA
921.54
USD
101884-892
101884-894
Ethyl-2-cyano-5-(trifluoromethyl)nicotinate ≥95%
Ethyl-2-cyano-5-(trifluoromethyl)nicotinate
Hazard: Irritant
Formula:
C₁₀H₇F₃N₂O₂ Melting Pt: 55…57 °C |
MDL Number:
MFCD13193103 CAS Number: 1192263-82-3 |
Specification Test Results
Melting point | 55-57°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...