Ethyl-4-bromo-5-(trifluoromethyl)-1H-pyrazole-3-carboxylate ≥97%

Supplier: Matrix Scientific

048690-500MG 048690-1G
101884-884EA 296.92 USD
101884-884 101884-886
Ethyl-4-bromo-5-(trifluoromethyl)-1H-pyrazole-3-carboxylate ≥97%
Ethyl-4-bromo-5-(trifluoromethyl)-1H-pyrazole-3-carboxylate

Hazard: Irritant

Formula: C₇H₆BrF₃N₂O₂
MW: 287.036 g/mol
Melting Pt: 131…133 °C
MDL Number: MFCD13193102
CAS Number: 1240278-21-0

Order Now

Specification Test Results

Melting point 131-133°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR