Ethyl-3-bromo-6-nitroimidazo[1,2-a]pyridine-2-carboxylate ≥95%

Supplier: Matrix Scientific

048689-500MG 048689-1G
101884-880EA 375.38 USD
101884-880 101884-882
Ethyl-3-bromo-6-nitroimidazo[1,2-a]pyridine-2-carboxylate ≥95%
Ethyl 3-bromo-6-nitroimidazo[1,2-a]pyridine-2-carboxylate

Hazard: Irritant

Formula: C₁₀H₈BrN₃O₄
Melting Pt: 173-175°C
MDL Number: MFCD13193101
CAS Number: 1198569-36-6

Order Now

Specification Test Results

Melting point 173-175°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR