Ethyl-5-amino-4,6-dichloronicotinate ≥95%

Supplier: Matrix Scientific

048687-500MG 048687-1G
101884-872EA 500 USD
101884-872 101884-874
Ethyl-5-amino-4,6-dichloronicotinate ≥95%
Ethyl-5-amino-4,6-dichloronicotinate

Hazard: Irritant

Formula: C₈H₈Cl₂N₂O₂
MW: 235.07 g/mol
Melting Pt: 75…77 °C
MDL Number: MFCD13193099
CAS Number: 154012-16-5

Order Now

Specification Test Results

Melting point 75-77°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR