4,6-Dichloro-5-nitronicotinic acid ≥95%
Supplier: Matrix Scientific
048683-500MG
048683-1G
101884-858EA
526.22
USD
101884-858
101884-860
4,6-Dichloro-5-nitronicotinic acid ≥95%
4,6-Dichloro-5-nitronicotinic acid
Hazard: Irritant
Formula:
C₆H₂Cl₂N₂O₄ Melting Pt: 217…220 °C |
MDL Number:
MFCD13193097 CAS Number: 1192263-83-4 |
Specification Test Results
Melting point | 217-220°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...