4,6-Dichloronicotinonitrile ≥95%

Supplier: Matrix Scientific

048682-500MG 048682-1G
101884-854EA 330.77 USD
101884-854 101884-856
4,6-Dichloronicotinonitrile ≥95%
4,6-Dichloronicotinonitrile

Hazard: Irritant

Formula: C₆H₂Cl₂N₂
Melting Pt: 133…135 °C
MDL Number: MFCD08276124
CAS Number: 166526-03-0

Order Now

Specification Test Results

Melting point 133-135°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR