2,3-Dichloroisonicotinamide ≥95%
Supplier: Matrix Scientific
048679-500MG
048679-1G
101884-844EA
443.08
USD
101884-844
101884-846
2,3-Dichloroisonicotinamide ≥95%
2,3-Dichloroisonicotinamide
Hazard: Irritant
Formula:
C₆H₄Cl₂N₂O Melting Pt: 171…173 °C |
MDL Number:
MFCD13193095 CAS Number: 1192263-98-1 |
Specification Test Results
Melting point | 171-173°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...