4-Chloro-1-methyl-7-nitro-1H-indazol-3-amine ≥95%

Supplier: Matrix Scientific

048675-500MG 048675-1G
101884-828EA 556.92 USD
101884-828 101884-830
4-Chloro-1-methyl-7-nitro-1H-indazol-3-amine ≥95%
4-Chloro-1-methyl-7-nitro-1H-indazol-3-amine

Hazard: Irritant

Formula: C₈H₇ClN₄O₂
Melting Pt: 186…189 °C
MDL Number: MFCD13193092
CAS Number: 1192263-90-3

Order Now

Specification Test Results

Melting point 186-189°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR