2-Chloro-5-methyl[1,6]naphthyridine ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Chloro-5-methyl-1,6-naphthyridine
048674-500MG
048674-1G
101884-824EA
570.77
USD
101884-824
101884-826
2-Chloro-5-methyl[1,6]naphthyridine ≥95%
2-Chloro-5-methyl[1,6]naphthyridine
Hazard: Irritant
Formula:
C₉H₇ClN₂ MW: 178.62 g/mol Melting Pt: 96…98 °C |
MDL Number:
MFCD01443634 CAS Number: 140692-93-9 |
Specification Test Results
Melting point | 96-98°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...