2-Chloro-6-cyclopropylnicotinonitrile ≥95%
Supplier: Matrix Scientific
048672-500MG
048672-1G
101884-814EA
356.92
USD
101884-814
101884-816
2-Chloro-6-cyclopropylnicotinonitrile ≥95%
2-Chloro-6-cyclopropylnicotinonitrile
Hazard: Irritant
Formula:
C₉H₇ClN₂ MW: 178.62 g/mol Melting Pt: 100…102 °C |
MDL Number:
MFCD13193091 CAS Number: 1198475-35-2 |
Specification Test Results
Melting point | 100-102°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...