4-Bromo-5-(trifluoromethyl)-1H-pyrazole-3-carboxylic acid ≥95%
Supplier: Matrix Scientific
048670-500MG
048670-1G
101884-808EA
747.69
USD
101884-808
101884-810
4-Bromo-5-(trifluoromethyl)-1H-pyrazole-3-carboxylic acid ≥95%
4-Bromo-5-(trifluoromethyl)-1H-pyrazole-3-carboxylic acid
Hazard: Irritant
Formula:
C₅H₂BrF₃N₂O₂ Melting Pt: 212…214 °C |
MDL Number:
MFCD13193089 CAS Number: 1257842-67-3 |
Specification Test Results
Melting point | 212-214°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...