3-Bromo-6-nitroimidazo[1,2-a]pyridine ≥95%
Supplier: Matrix Scientific
048669-500MG
048669-1G
101884-804EA
149.23
USD
101884-804
101884-806
3-Bromo-6-nitroimidazo[1,2-a]pyridine ≥95%
3-Bromo-6-nitroimidazo[1,2-a]pyridine
Hazard: Irritant
Formula:
C₇H₄BrN₃O₂ MW: 242.0296 g/mol Melting Pt: 140-142°C |
MDL Number:
MFCD13193088 CAS Number: 52310-42-6 |
Specification Test Results
Melting point | 140-142°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...