4-Bromo-7-methoxy-1H-pyrrolo[2,3-c]pyridine ≥95%
Supplier: Matrix Scientific
048667-500MG
048667-1G
101884-798EA
600
USD
101884-798
101884-800
4-Bromo-7-methoxy-1H-pyrrolo[2,3-c]pyridine ≥95%
4-Bromo-7-methoxy-1H-pyrrolo[2,3-c]pyridine
Hazard: Irritant
Formula:
C₈H₇BrN₂O Melting Pt: 174…175 °C |
MDL Number:
MFCD11845544 CAS Number: 425380-37-6 |
Specification Test Results
Melting point | 174-175°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...