3-Amino-5-bromo-2-methoxypyridine ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Bromo-2-methoxypyridin-3-amine
048666-1G
048666-5G
101884-794EA
15.38
USD
101884-794
101884-796
3-Amino-5-bromo-2-methoxypyridine ≥95%
3-Amino-5-bromo-2-methoxypyridine
Hazard: Irritant
Formula:
C₆H₇BrN₂O Melting Pt: 57-58°C |
MDL Number:
MFCD07368887 CAS Number: 884495-39-0 |
Specification Test Results
Melting point | 57-58°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...