6-Bromo-5-methoxy-2-pyridinecarbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Bromo-5-methoxypicolinaldehyde

048665-500MG 048665-1G
101884-790EA 355.38 USD
101884-790 101884-792
6-Bromo-5-methoxy-2-pyridinecarbaldehyde ≥95%
6-Bromo-5-methoxy-2-pyridinecarbaldehyde

Hazard: Irritant

Formula: C₇H₆BrNO₂
MW: 216.04 g/mol
Melting Pt: 122…124 °C
MDL Number: MFCD13193086
CAS Number: 329217-74-5

Order Now

Specification Test Results

Melting point 122-124°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR