4-Bromo-2-iodo-5-(trifluoromethyl)aniline ≥97%
Supplier: Matrix Scientific
048664-500MG
048664-1G
048664-5G
101884-784EA
278.46
USD
101884-784
101884-786
101884-788
4-Bromo-2-iodo-5-(trifluoromethyl)aniline ≥97%
4-Bromo-2-iodo-5-(trifluoromethyl)aniline
Hazard: Irritant
Formula:
C₇H₄BrF₃IN MW: 365.9170 g/mol Melting Pt: 64-66°C |
MDL Number:
MFCD13193085 CAS Number: 868692-81-3 |
Specification Test Results
Melting point | 64-66°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...