3-Bromo-5-hydroxy-2(1H)-quinolinone ≥93%

Supplier: Matrix Scientific

048663-500MG 048663-1G
101884-780EA 1109.23 USD
101884-780 101884-782
3-Bromo-5-hydroxy-2(1H)-quinolinone ≥93%
3-Bromo-5-hydroxy-2(1H)-quinolinone

Hazard: Irritant

Formula: C₉H₆BrNO₂
MW: 240.06 g/mol
Melting Pt: >300 °C
MDL Number: MFCD13193084
CAS Number: 1192263-85-6

Order Now

Specification Test Results

Melting point >300°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR