4-Bromo-N,2-dihydroxybenzenecarboxamide ≥95%

Supplier: Matrix Scientific

048661-500MG 048661-1G
101884-772EA 529.23 USD
101884-772 101884-774
4-Bromo-N,2-dihydroxybenzenecarboxamide ≥95%
4-Bromo-N,2-dihydroxybenzenecarboxamide

Hazard: Irritant

Formula: C₇H₆BrNO₃
MW: 232.04 g/mol
Melting Pt: 225…226 °C (dec.)
MDL Number: MFCD13193083
CAS Number: 61799-79-9

Order Now

Specification Test Results

Melting point 225-226(de


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR