2-Bromo-5-chloro-4-(trifluoromethyl)aniline ≥95%
Supplier: Matrix Scientific
048660-500MG
048660-1G
101884-768EA
747.69
USD
101884-768
101884-770
2-Bromo-5-chloro-4-(trifluoromethyl)aniline ≥95%
2-Bromo-5-chloro-4-(trifluoromethyl)aniline
Hazard: Irritant
Formula:
C₇H₄BrClF₃N MW: 274.4700 g/mol Melting Pt: 31…33 °C |
MDL Number:
MFCD13193082 CAS Number: 863111-48-2 |
Specification Test Results
Melting point | 31-33°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...