4-Bromo-7-chloro-1H-pyrrolo[2,3-c]pyridine ≥95%

Supplier: Matrix Scientific

048658-500MG 048658-1G
101884-760EA 600 USD
101884-760 101884-762
4-Bromo-7-chloro-1H-pyrrolo[2,3-c]pyridine ≥95%
4-Bromo-7-chloro-1H-pyrrolo[2,3-c]pyridine

Hazard: Irritant

Formula: C₇H₄BrClN₂
Melting Pt: 230…231 °C
MDL Number: MFCD11848529
CAS Number: 425380-38-7

Order Now

Specification Test Results

Melting point 230-231°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR