4-Bromo-6-chloro-3-phenylpyridazine ≥95%
Supplier: Matrix Scientific
048657-500MG
048657-1G
101884-756EA
523.08
USD
101884-756
101884-758
4-Bromo-6-chloro-3-phenylpyridazine ≥95%
4-Bromo-6-chloro-3-phenylpyridazine
Hazard: Irritant
Formula:
C₁₀H₆BrClN₂ Melting Pt: 84…86 °C |
MDL Number:
MFCD13193081 CAS Number: 433935-99-0 |
Specification Test Results
Melting point | 84-86°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...