1-Bromo-4-chloro-2-nitro-5-(trifluoromethyl)benzene ≥95%

Supplier: Matrix Scientific

048656-500MG 048656-1G
101884-752EA 529.23 USD
101884-752 101884-754
1-Bromo-4-chloro-2-nitro-5-(trifluoromethyl)benzene ≥95%
1-Bromo-4-chloro-2-nitro-5-(trifluoromethyl)benzene

Hazard: Irritant

Formula: C₇H₂BrClF₃NO₂
Melting Pt: 58…60 °C
MDL Number: MFCD13193080
CAS Number: 863111-47-1

Order Now

Specification Test Results

Melting point 58-60°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR