6-Bromo-1,2-benzisoxazol-3-ol ≥90%
Supplier: Matrix Scientific
Synonyms:
6-Bromobenzo[d]isoxazol-3(2H)-one
048654-500MG
048654-1G
101884-746EA
921.54
USD
101884-746
101884-748
6-Bromo-1,2-benzisoxazol-3-ol ≥90%
6-Bromo-1,2-benzisoxazol-3-ol
Hazard: Irritant
Formula:
C₇H₄BrNO₂ MW: 214.02 g/mol Melting Pt: 219…221 °C |
MDL Number:
MFCD13193078 CAS Number: 65685-51-0 |
Specification Test Results
Melting point | 219-221°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...