5-(Benzyloxy)-6-chloro-2-pyridinecarbaldehyde ≥95%

Supplier: Matrix Scientific

048651-500MG 048651-1G
101884-734EA 623.08 USD
101884-734 101884-736
5-(Benzyloxy)-6-chloro-2-pyridinecarbaldehyde ≥95%
5-(Benzyloxy)-6-chloro-2-pyridinecarbaldehyde

Hazard: Irritant

Formula: C₁₃H₁₀ClNO₂
Melting Pt: 105…107 °C
MDL Number: MFCD13193075
CAS Number: 59781-07-6

Order Now

Specification Test Results

Melting point 105-107°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR