3-(Benzyloxy)-2-chloro-6-(chloromethyl)-4-iodopyridine ≥95%
Supplier: Matrix Scientific
048649-500MG
048649-1G
101884-726EA
921.54
USD
101884-726
101884-728
3-(Benzyloxy)-2-chloro-6-(chloromethyl)-4-iodopyridine ≥95%
3-(Benzyloxy)-2-chloro-6-(chloromethyl)-4-iodopyridine
Hazard: Irritant
Formula:
C₁₃H₁₀Cl₂INO Melting Pt: 114…116 °C |
MDL Number:
MFCD13193073 CAS Number: 1192263-74-3 |
Specification Test Results
Melting point | 114-116°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...